(S,S)-N,N'-Bis(1-hydroxy-2-butyl)ethylenediamine dihydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
Ethambutol Hydrochloride, (2S,2'S)-2,2'-(Ethane-1,2-diylbis(azanediyl))bis(butan-1-ol) dihydrochloride, (S,S)-2,2'-(1,2-Ethanediyldiimino)bis-1-butanol Dihydrochloride
CAS Number: 1070-11-7
MDL Number: MFCD00216025
Molecular Formula: C10H24N2O2
Molecular Weight: 277.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 199
Specific rotation [a]20/D: 6 deg (C=10, H2O)
Product Categories: Bioscience, Synthetic Antimicrobial Ingredients
Other Product Names / Synonyms: (S,S)-2,2'-(1,2-Ethanediyldiimino)bis-1-butanol Dihydrochloride
Formula:
C₁₀H₂₄N₂O₂·2HCl MW: 277.23 g/mol Melting Pt: 199 °C |
MDL Number:
MFCD00216025 CAS Number: 1070-11-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...