Ethyl-2-amino-4,5-dimethylthiophene-3-carboxylate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
Ethyl 2-amino-4,5-dimethyl-3-thiophenecarboxylate, Ethyl 2-amino-4,5-dimethylthiophene-3-carboxylate
E1005-25G
E1005-5G
TCE1005-25GEA
1594.22
CAD
TCE1005-25G
TCE1005-5G
Ethyl-2-amino-4,5-dimethylthiophene-3-carboxylate ≥98.0% (by GC, titration analysis)
Ethyl 2-amino-4,5-dimethylthiophene-3-carboxylate
CAS Number: 4815-24-1
MDL Number: MFCD00085051
Molecular Formula: C9H13NO2S
Molecular Weight: 199.27
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 93
Lambda max.: 310 nm (MeOH)
Other Product Names / Synonyms: 2-Amino-4,5-dimethylthiophene-3-carboxylic Acid Ethyl Ester
Formula:
C₉H₁₃NO₂S MW: 199.27 g/mol Melting Pt: 93 °C |
MDL Number:
MFCD00085051 CAS Number: 4815-24-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...