Ethyl-1,5-Dimethylpyrazole-3-carboxylate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Ethyl 1,5-dimethyl-1H-pyrazole-3-carboxylate, 1,5-Dimethylpyrazole-3-carboxylic Acid Ethyl Ester

E1002-1G E1002-200MG
TCE1002-1GEA 171.9 CAD
TCE1002-1G TCE1002-200MG
Ethyl-1,5-Dimethylpyrazole-3-carboxylate ≥98.0% (by GC)
Ethyl-1,5-Dimethylpyrazole-3-carboxylate

CAS Number: 5744-51-4
MDL Number: MFCD00085071
Molecular Formula: C8H12N2O2
Molecular Weight: 168.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 119
Melting point (°C): 45


Other Product Names / Synonyms: 1,5-Dimethylpyrazole-3-carboxylic Acid Ethyl Ester

Formula: C₈H₁₂N₂O₂
MW: 168.20 g/mol
Boiling Pt: 119 °C
Melting Pt: 45 °C
MDL Number: MFCD00085071
CAS Number: 5744-51-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bcfe3104235099a016a431a4770e157ff45ac8e5&vid=bcfe3104235099a016a431a4770e157ff45ac8e5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR