Ethyl-5-amino-1-methyl-1H-pyrazole-4-carboxylate ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 5-amino-1-methyl-1H-pyrazole-4-carboxylate

E0986-25G E0986-5G
TCE0986-25GEA 115.34 CAD
TCE0986-25G TCE0986-5G
Ethyl-5-amino-1-methyl-1H-pyrazole-4-carboxylate ≥98.0%
Ethyl 5-amino-1-methyl-1H-pyrazole-4-carboxylate

CAS Number: 31037-02-2
MDL Number: MFCD00051652
Molecular Formula: C7H11N3O2
Molecular Weight: 169.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 101
Lambda max.: 257 nm (EtOH)


Other Product Names / Synonyms: 5-Amino-1-methylpyrazole-4-carboxylic Acid Ethyl Ester

Formula: C₇H₁₁N₃O₂
Melting Pt: 101 °C
MDL Number: MFCD00051652
CAS Number: 31037-02-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2391494b16fe9927cdc2156b58d2283f008ac357&vid=2391494b16fe9927cdc2156b58d2283f008ac357&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR