Ethyl-2-aminobenzo[d]thiazole-6-carboxylate ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 2-aminobenzo[d]thiazole-6-carboxylate

E0985-1G E0985-5G
TCE0985-1GEA 44.56 CAD
TCE0985-1G TCE0985-5G
Ethyl-2-aminobenzo[d]thiazole-6-carboxylate ≥98.0%
Ethyl-2-aminobenzo[d]thiazole-6-carboxylate

CAS Number: 50850-93-6
MDL Number: MFCD00102724
Molecular Formula: C10H10N2O2S
Molecular Weight: 222.26
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 242


Other Product Names / Synonyms: 2-Aminobenzothiazole-6-carboxylic Acid Ethyl Ester

Formula: C₁₀H₁₀N₂O₂S
MW: 222.26 g/mol
Melting Pt: 242 °C
MDL Number: MFCD00102724
CAS Number: 50850-93-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41e90c87ff227dfe70abac6a7363e1fae1f543d7&vid=41e90c87ff227dfe70abac6a7363e1fae1f543d7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR