1-tert-Butyl-3-ethyl-4-oxopiperidine-1,3-dicarboxylate ≥97.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 1-tert-Butoxycarbonyl-4-oxo-3-piperidinecarboxylic acid ethyl ester, Ethyl 1-Boc-4-oxo-3-piperidinecarboxylate, Ethyl 1-tert-Butoxycarbonyl-4-oxo-3-piperidinecarboxylate, 1-Boc-3-ethoxycarbonyl-4-piperidone, 1-tert-Butyl 3-ethyl 4-oxopiperidine-1,3-dicarboxylate, 1-Boc-4-oxo-3-piperidinecarboxylic acid ethyl ester, 1-tert-Butoxycarbonyl-3-ethoxycarbonyl-4-piperidone

E0983-1G E0983-5G
TCE0983-1GEA 64.78 CAD
TCE0983-1G TCE0983-5G
1-tert-Butyl-3-ethyl-4-oxopiperidine-1,3-dicarboxylate ≥97.0% (by HPLC, total nitrogen)
1-tert-Butyl-3-ethyl-4-oxopiperidine-1,3-dicarboxylate

CAS Number: 98977-34-5
MDL Number: MFCD01862189
Molecular Formula: C13H21NO5
Molecular Weight: 271.31
Purity/Analysis Method: >97.0% (HPLC,N)
Form: Crystal
Melting point (°C): 64


Product Categories: Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)

Formula: C13H21NO5
MW: 271.3100 g/mol
Melting Pt: 64 °C
MDL Number: MFCD01862189
CAS Number: 98977-34-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70724325b4d088c4090791ca72b78fd065bc0a88&vid=70724325b4d088c4090791ca72b78fd065bc0a88&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR