Ethyl-5-amino-4-cyano-3-(2-ethoxycarbonylmethyl)thiophene-2-carboxylate 98,0

Supplier: TCI America

Synonyms: 5-Amino-4-cyano-3-(2-ethoxycarbonylmethyl)thiophene-2-carboxylic Acid Ethyl Ester, Ethyl 5-amino-4-cyano-3-(2-ethoxy-2-oxoethyl)thiophene-2-carboxylate

E0980-25G E0980-5G
TCE0980-25GEA 107.44 CAD
TCE0980-25G TCE0980-5G
Ethyl-5-amino-4-cyano-3-(2-ethoxycarbonylmethyl)thiophene-2-carboxylate 98,0
Ethyl-5-amino-4-cyano-3-(2-ethoxycarbonylmethyl)thiophene-2-carboxylate

CAS Number: 58168-20-0
MDL Number: MFCD00832845
Molecular Formula: C12H14N2O4S
Molecular Weight: 282.31
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 139


Other Product Names / Synonyms: 5-Amino-4-cyano-3-(2-ethoxycarbonylmethyl)thiophene-2-carboxylic Acid Ethyl Ester

Formula: C₁₂H₁₄N₂O₄S
MW: 282.31 g/mol
Melting Pt: 139 °C
MDL Number: MFCD00832845
CAS Number: 58168-20-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9ede797bbad8b7631e8e1b835530ee8359609f7&vid=c9ede797bbad8b7631e8e1b835530ee8359609f7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR