3-(2-Ethylhexyl)thiophene ≥97.0% (by GC)

Supplier: TCI America

E0973-1G E0973-200MG
TCE0973-1GEA 666.76 CAD
TCE0973-1G TCE0973-200MG
3-(2-Ethylhexyl)thiophene ≥97.0% (by GC)
3-(2-Ethylhexyl)thiophene

CAS Number: 121134-38-1
MDL Number: MFCD02260336
Molecular Formula: C12H20S
Molecular Weight: 196.35
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 30
Storage Temperature: 0-10°C

Formula: C₁₂H₂₀S
Boiling Pt: 30 °C
MDL Number: MFCD02260336
CAS Number: 121134-38-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99b86306f4cd3456e9a07bc6708eba036a42f9ab&vid=99b86306f4cd3456e9a07bc6708eba036a42f9ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR