1-Ethynyl-4-(phenylethynyl)benzene ≥98.0% (by GC)

Supplier: TCI America

E0967-1G E0967-200MG
TCE0967-1GEA 816.54 CAD
TCE0967-1G TCE0967-200MG
1-Ethynyl-4-(phenylethynyl)benzene ≥98.0% (by GC)
1-Ethynyl-4-(phenylethynyl)benzene

CAS Number: 92866-00-7
Molecular Formula: C16H10
Molecular Weight: 202.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 90
Lambda max.: 318 nm (Benzene)


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: 4-Ethynyldiphenylacetylene, 1-(4-Ethynylphenyl)-2-phenylacetylene

Formula: C₁₆H₁₀
Melting Pt: 90 °C
MDL Number: MFCD27665710
CAS Number: 92866-00-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1a5596cf71165fca5c2a4a2fc888ca00cd6b11b&vid=d1a5596cf71165fca5c2a4a2fc888ca00cd6b11b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR