Ethyl pyrazole-3-carboxylate ≥98.0% (by GC)

Supplier: TCI America

E0965-1G E0965-5G
TCE0965-1GEA 33.18 CAD
TCE0965-1G TCE0965-5G
Ethyl pyrazole-3-carboxylate ≥98.0% (by GC)
Ethyl pyrazole-3-carboxylate

CAS Number: 5932-27-4
MDL Number: MFCD00159643
Molecular Formula: C6H8N2O2
Molecular Weight: 140.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 161


Other Product Names / Synonyms: Pyrazole-3-carboxylic Acid Ethyl Ester

Formula: C₆H₈N₂O₂
Melting Pt: 161 °C
MDL Number: MFCD00159643
CAS Number: 5932-27-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec5c4bad3a2f0ab3e61ed4fb6385dfbc45f36615&vid=ec5c4bad3a2f0ab3e61ed4fb6385dfbc45f36615&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR