3-Ethyl-4-methyl-3-pyrrolin-2-one ≥98.0%

Supplier: TCI America

Synonyms: 3-Ethyl-4-methyl-2,5-dihydro-1H-pyrrol-2-one, 1,5-Dihydro-3-ethyl-4-methyl-2H-pyrrol-2-one

E0964-25G E0964-5G
TCE0964-25GEA 556.16 CAD
TCE0964-25G TCE0964-5G
3-Ethyl-4-methyl-3-pyrrolin-2-one ≥98.0%
3-Ethyl-4-methyl-3-pyrrolin-2-one

CAS Number: 766-36-9
MDL Number: MFCD00173861
Molecular Formula: C7H11NO
Molecular Weight: 125.17
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 104

Formula: C₇H₁₁NO
MW: 125.17 g/mol
Melting Pt: 104 °C
MDL Number: MFCD00173861
CAS Number: 766-36-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58f2dfc16f1dd46ecbd1ffdf30610b316715df52&vid=58f2dfc16f1dd46ecbd1ffdf30610b316715df52&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR