Econazole nitrate ≥98.0%

Supplier: TCI America

Synonyms: 1-[2-(4-Chlorobenzyloxy)-2-(2,4-dichlorophenyl)ethyl]imidazole Nitrate, Ecostatin, 1-[2-(4-Chlorophenylmethoxy)-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole nitrate

E0957-25G E0957-5G
TCE0957-25GEA 312.84 CAD
TCE0957-25G TCE0957-5G
Econazole nitrate ≥98.0%
Econazole nitrate

CAS Number: 24169-02-6
MDL Number: MFCD00058160
Molecular Formula: C18H15Cl3N2O
Molecular Weight: 444.69
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 161


Product Categories: Bioscience, Antifungal Ingredients
Other Product Names / Synonyms: 1-[2-(4-Chlorobenzyloxy)-2-(2,4-dichlorophenyl)ethyl]imidazole Nitrate

Formula: C₁₈H₁₅Cl₃N₂O
MW: 444.69 g/mol
Melting Pt: 161 °C
MDL Number: MFCD00058160
CAS Number: 24169-02-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1cc3ea5efc7c274f60b2a9f8b803a9db768ab7f&vid=e1cc3ea5efc7c274f60b2a9f8b803a9db768ab7f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR