Ethyl indole-3-carboxylate ≥97.0%

Supplier: TCI America

Synonyms: Indole-3-carboxylic acid ethyl ester, Ethyl 1H-indole-3-carboxylate

E0955-5G E0955-1G
TCE0955-5GEA 135.56 CAD
TCE0955-5G TCE0955-1G
Ethyl indole-3-carboxylate ≥97.0%
Ethyl indole-3-carboxylate

CAS Number: 776-41-0
MDL Number: MFCD00228454
Molecular Formula: C11H11NO2
Molecular Weight: 189.21
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 123


Other Product Names / Synonyms: Indole-3-carboxylic Acid Ethyl Ester

Formula: C₁₁H₁₁NO₂
MW: 189.21 g/mol
Melting Pt: 123 °C
MDL Number: MFCD00228454
CAS Number: 776-41-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a085ae7b62e6965cc0b905cd86ab2b1d40a12177&vid=a085ae7b62e6965cc0b905cd86ab2b1d40a12177&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR