1-Boc-DL-nipecotic acid ethyl ester ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 1-(tert-Butoxycarbonyl)-3-piperidinecarboxylate, Ethyl 1-Boc-DL-nipecotate, Ethyl 1-Boc-3-piperidinecarboxylate

E0954-25G E0954-5G
TCE0954-25GEA 153.73 CAD
TCE0954-25G TCE0954-5G
1-Boc-DL-nipecotic acid ethyl ester ≥98.0%
1-Boc-DL-nipecotic acid ethyl ester

CAS Number: 130250-54-3
MDL Number: MFCD04116274
Molecular Formula: C13H23NO4
Molecular Weight: 257.33
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 93
Melting point (°C): 37


Product Categories: Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)

Formula: C₁₃H₂₃NO₄
MW: 257.33 g/mol
Boiling Pt: 93 °C
Melting Pt: 37 °C
MDL Number: MFCD04116274
CAS Number: 130250-54-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72abe3039ca7fdae51c7f62f65bda51706542bae&vid=72abe3039ca7fdae51c7f62f65bda51706542bae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR