Imidazole-2-carboxylic acid ethyl ester ≥98.0%

Supplier: TCI America

Synonyms: Ethyl imidazole-2-carboxylate, Ethyl 1H-imidazole-2-carboxylate

E0952-1G E0952-5G
TCE0952-1GEA 105.86 CAD
TCE0952-1G TCE0952-5G
Imidazole-2-carboxylic acid ethyl ester ≥98.0%
Imidazole-2-carboxylic acid ethyl ester

CAS Number: 33543-78-1
MDL Number: MFCD03426031
Molecular Formula: C6H8N2O2
Molecular Weight: 140.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 51
Melting point (°C): 178


Other Product Names / Synonyms: 2-Imidazolecarboxylic Acid Ethyl Ester

Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
Boiling Pt: 51 °C
Melting Pt: 178 °C
MDL Number: MFCD03426031
CAS Number: 33543-78-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85bfa139ecee0cb272bc53858822b7a0f8c18c68&vid=85bfa139ecee0cb272bc53858822b7a0f8c18c68&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR