Ethyl-5-nitrobenzofuran-2-carboxylate ≥97.0% (by GC)

Supplier: TCI America

E0950-25G E0950-5G
TCE0950-25GEA 243 CAD
TCE0950-25G TCE0950-5G
Ethyl-5-nitrobenzofuran-2-carboxylate ≥97.0% (by GC)
Ethyl-5-nitrobenzofuran-2-carboxylate

CAS Number: 69604-00-8
MDL Number: MFCD00275272
Molecular Formula: C11H9NO5
Molecular Weight: 235.20
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 153


Other Product Names / Synonyms: 5-Nitrobenzofuran-2-carboxylic Acid Ethyl Ester

Formula: C₁₁H₉NO₅
Melting Pt: 153 °C
MDL Number: MFCD00275272
CAS Number: 69604-00-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6399dae0bd110cb2b75befd382c2eab384bc3c6e&vid=6399dae0bd110cb2b75befd382c2eab384bc3c6e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR