Ethyl-2-bromo-4-methylthiazole-5-carboxylate ≥97.0%
Supplier: TCI America
Synonyms:
Ethyl 2-bromo-4-methylthiazole-5-carboxylate
E0945-1G
E0945-5G
TCE0945-1GEA
67.78
CAD
TCE0945-1G
TCE0945-5G
Ethyl-2-bromo-4-methylthiazole-5-carboxylate ≥97.0%
Ethyl-2-bromo-4-methylthiazole-5-carboxylate
CAS Number: 22900-83-0
MDL Number: MFCD03791227
Molecular Formula: C7H8BrNO2S
Molecular Weight: 250.11
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 68
Other Product Names / Synonyms: 2-Bromo-4-methylthiazole-5-carboxylic Acid Ethyl Ester
Formula:
C₇H₈BrNO₂S Melting Pt: 68 °C |
MDL Number:
MFCD03791227 CAS Number: 22900-83-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...