Ethyl-2-aminoisonicotinate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

E0944-1G E0944-5G
TCE0944-1GEA 52.14 CAD
TCE0944-1G TCE0944-5G
Ethyl-2-aminoisonicotinate ≥98.0% (by GC, titration analysis)
Ethyl 2-aminoisonicotinate

CAS Number: 13362-30-6
MDL Number: MFCD03791260
Molecular Formula: C8H10N2O2
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 125


Other Product Names / Synonyms: 2-Aminoisonicotinic Acid Ethyl Ester, Ethyl 2-Aminopyridine-4-carboxylate, 2-Aminopyridine-4-carboxylic Acid Ethyl Ester

Formula: C₈H₁₀N₂O₂
Melting Pt: 125 °C
MDL Number: MFCD03791260
CAS Number: 13362-30-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=751a2f5e88753fc55b52fd45478e1c3498b238ac&vid=751a2f5e88753fc55b52fd45478e1c3498b238ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR