Ethyl-2-oxocyclopentaneacetate ≥95.0%
Supplier: TCI America
Synonyms:
2-Oxocyclopentaneacetic Acid Ethyl Ester, Ethyl 2-oxocyclopentylacetate, Ethyl 2-(2-Oxocyclopentyl)acetate, 2-(2-Oxocyclopentyl)acetic Acid Ethyl Ester, Ethyl 2-(2-oxocyclopentyl)acetate
CAS Number: 20826-94-2
MDL Number: MFCD00044715
Molecular Formula: C9H14O3
Molecular Weight: 170.21
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 106
Specific Gravity (20/20): 1.06
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 2-Oxocyclopentaneacetic Acid Ethyl Ester, Ethyl 2-(2-Oxocyclopentyl)acetate, 2-(2-Oxocyclopentyl)acetic Acid Ethyl Ester
Formula:
C₉H₁₄O₃ MW: 170.21 g/mol Boiling Pt: 106 °C Density: 1.06 (20 °C) |
MDL Number:
MFCD00044715 CAS Number: 20826-94-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...