1-Ethynylpyrene ≥98.0% (by GC)

Supplier: TCI America

E0939-1G E0939-200MG
TCE0939-1GEA 1520.75 CAD
TCE0939-1G TCE0939-200MG
1-Ethynylpyrene ≥98.0% (by GC)
1-Ethynylpyrene

CAS Number: 34993-56-1
MDL Number: MFCD02093933
Molecular Formula: C18H10
Molecular Weight: 226.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 117

Formula: C₁₈H₁₀
Melting Pt: 117 °C
MDL Number: MFCD02093933
CAS Number: 34993-56-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c72f2ce6bd64943805c95220369d4f4ee4a44d7&vid=8c72f2ce6bd64943805c95220369d4f4ee4a44d7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR