Ethyl-1-aminocyclopropanecarboxylate hydrochloride 98,0

Supplier: TCI America

Synonyms: H-Acpc-OEt.HCl, H-Acpc-OEt·HCl

E0937-1G E0937-5G
TCE0937-1GEA 50.56 CAD
TCE0937-1G TCE0937-5G
Ethyl-1-aminocyclopropanecarboxylate hydrochloride 98,0
Ethyl-1-aminocyclopropanecarboxylate hydrochloride

CAS Number: 42303-42-4
MDL Number: MFCD00190747
Molecular Formula: C6H11NO2
Molecular Weight: 165.62
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 110


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: 1-Aminocyclopropanecarboxylic Acid Ethyl Ester Hydrochloride

Formula: C₆H₁₁NO₂·HCl
MW: 165.6 g/mol
Melting Pt: 110 °C
MDL Number: MFCD00190747
CAS Number: 42303-42-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=490c1f736746f6689c2c1cd692e8c42fb9aab22c&vid=490c1f736746f6689c2c1cd692e8c42fb9aab22c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR