Ethyl-2,4-difluorobenzoate ≥98.0% (by GC)
Supplier: TCI America
E0935-25G
E0935-5G
TCE0935-25GEA
118.97
CAD
TCE0935-25G
TCE0935-5G
Ethyl-2,4-difluorobenzoate ≥98.0% (by GC)
Ethyl 2,4-difluorobenzoate
CAS Number: 108928-00-3
MDL Number: MFCD00153149
Molecular Formula: C9H8F2O2
Molecular Weight: 186.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 88
Specific Gravity (20/20): 1.24
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2,4-Difluorobenzoic Acid Ethyl Ester
Formula:
C₉H₈F₂O₂ Boiling Pt: 88 °C Density: 1.24 (20 °C) |
MDL Number:
MFCD00153149 CAS Number: 108928-00-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...