Ethyl-2-(2-methyl-1,3-dioxolan-2-yl)acetate ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 2-(2-methyl-1,3-dioxolan-2-yl)acetate, 2-(2-Methyl-1,3-dioxolan-2-yl)acetic Acid Ethyl Ester

E0929-25G E0929-500G E0929-100G
TCE0929-25GEA 104.28 CAD
TCE0929-25G TCE0929-500G TCE0929-100G
Ethyl-2-(2-methyl-1,3-dioxolan-2-yl)acetate ≥98.0%
Ethyl-2-(2-methyl-1,3-dioxolan-2-yl)acetate

CAS Number: 6413-10-1
MDL Number: MFCD00152488
Molecular Formula: C8H14O4
Molecular Weight: 174.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 98
Specific Gravity (20/20): 1.09


Other Product Names / Synonyms: 2-(2-Methyl-1,3-dioxolan-2-yl)acetic Acid Ethyl Ester

Formula: C₈H₁₄O₄
MW: 174.20 g/mol
Boiling Pt: 98 °C
Density: 1.09 (20 °C)
MDL Number: MFCD00152488
CAS Number: 6413-10-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ae8e67e56dc6171cac4990e6af27285cbae8b40&vid=1ae8e67e56dc6171cac4990e6af27285cbae8b40&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR