2-(3-Ethoxy-4-ethoxycarbonylphenyl)acetic acid ≥98.0%

Supplier: TCI America

Synonyms: 4-Carboxymethyl-2-ethoxybenzoic Acid Ethyl Ester, Ethyl 4-Carboxymethyl-2-ethoxybenzoate, 2-(3-Ethoxy-4-(ethoxycarbonyl)phenyl)acetic acid

E0928-25G E0928-5G
TCE0928-25GEA 502.44 CAD
TCE0928-25G TCE0928-5G
2-(3-Ethoxy-4-ethoxycarbonylphenyl)acetic acid ≥98.0%
2-(3-Ethoxy-4-ethoxycarbonylphenyl)acetic acid

CAS Number: 99469-99-5
MDL Number: MFCD08704233
Molecular Formula: C13H16O5
Molecular Weight: 252.27
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 80


Other Product Names / Synonyms: Ethyl 4-Carboxymethyl-2-ethoxybenzoate, 4-Carboxymethyl-2-ethoxybenzoic Acid Ethyl Ester

Formula: C₁₃H₁₆O₅
MW: 252.27 g/mol
Melting Pt: 80 °C
MDL Number: MFCD08704233
CAS Number: 99469-99-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49b435fefc875e0e5d3d34219dc2310efa7bed7c&vid=49b435fefc875e0e5d3d34219dc2310efa7bed7c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR