Ethyl-3-bromo-1H-indole-2-carboxylate ≥95.0% (by GC)

Supplier: TCI America

E0927-1G E0927-5G
TCE0927-1GEA 61.62 CAD
TCE0927-1G TCE0927-5G
Ethyl-3-bromo-1H-indole-2-carboxylate ≥95.0% (by GC)
Ethyl-3-bromo-1H-indole-2-carboxylate

CAS Number: 91348-45-7
MDL Number: MFCD02071793
Molecular Formula: C11H10BrNO2
Molecular Weight: 268.11
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 151


Other Product Names / Synonyms: 3-Bromoindole-2-carboxylic Acid Ethyl Ester

Formula: C₁₁H₁₀BrNO₂
Melting Pt: 151 °C
MDL Number: MFCD02071793
CAS Number: 91348-45-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74029adfa8e65306a89529aab13c29413bb407b1&vid=74029adfa8e65306a89529aab13c29413bb407b1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR