Ebastine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

E0925-1G E0925-5G
TCE0925-1GEA 218.04 CAD
TCE0925-1G TCE0925-5G
Ebastine ≥98.0% (by HPLC, titration analysis)
Ebastine

CAS Number: 90729-43-4
MDL Number: MFCD00865661
Molecular Formula: C32H39NO2
Molecular Weight: 469.67
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 87

Formula: C₃₂H₃₉NO₂
Melting Pt: 87 °C
MDL Number: MFCD00865661
CAS Number: 90729-43-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e212e6dd91b748fe2ecba9074babffba1293b0d&vid=4e212e6dd91b748fe2ecba9074babffba1293b0d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR