Ethyl-4-aminobutyrate hydrochloride ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America

E0924-25G E0924-5G
TCE0924-25GEA 121.66 CAD
TCE0924-25G TCE0924-5G
Ethyl-4-aminobutyrate hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Ethyl-4-aminobutyrate hydrochloride

CAS Number: 6937-16-2
MDL Number: MFCD00012915
Molecular Formula: C6H13NO2
Molecular Weight: 167.63
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 88


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: 4-Aminobutyric Acid Ethyl Ester Hydrochloride, H-GABA-OEt.HCl

Formula: C₆H₁₃NO₂
Melting Pt: 88 °C
MDL Number: MFCD00012915
CAS Number: 6937-16-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa251017ee603fdee57f3648526ca607162fe4ad&vid=aa251017ee603fdee57f3648526ca607162fe4ad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR