D-(-)-N-Ethylglucamine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: N-Ethyl-D-glucamine, (-)-1-Deoxy-1-(ethylamino)-D-glucitol, (-)-1-Ethylamino-1-deoxy-D-glucitol, (2R,3R,4R,5S)-6-(Ethylamino)hexane-1,2,3,4,5-pentaol, (-)-N-Ethyl-D-glucamine

E0923-250G E0923-25G
TCE0923-250GEA 116.92 CAD
TCE0923-250G TCE0923-25G
D-(-)-N-Ethylglucamine ≥98.0% (by titrimetric analysis)
D(-)-N-Ethylglucamine

CAS Number: 14216-22-9
MDL Number: MFCD03789564
Molecular Formula: C8H19NO5
Molecular Weight: 209.24
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 139
Specific rotation [a]20/D: -16 deg (C=1, H2O)

Formula: H₃CCH₂NHCH₂(CH(OH))₄CH₂OH
MW: 209.24 g/mol
Melting Pt: 139 °C
MDL Number: MFCD03789564
CAS Number: 14216-22-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf8452b324baf6ea25f0ddb88d7edb967fc280bb&vid=bf8452b324baf6ea25f0ddb88d7edb967fc280bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR