Ethylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

E0913-1G E0913-5G
TCE0913-1GEA 69.52 CAD
TCE0913-1G TCE0913-5G
Ethylboronic acid (contains varying amounts of Anhydride)
Ethylboronic acid

CAS Number: 4433-63-0
MDL Number: MFCD01074536
Molecular Formula: C2H7BO2
Molecular Weight: 73.89
Form: Crystal
Color: White


Other Product Names / Synonyms: Ethaneboronic Acid

Formula: C₂H₇BO₂
MDL Number: MFCD01074536
CAS Number: 4433-63-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=028875f4bfcecc1e79058e9d23cc46916c4e8590&vid=028875f4bfcecc1e79058e9d23cc46916c4e8590&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR