Ethyl-2-aminothiazole-5-carboxylate ≥97.0%

Supplier: TCI America

Synonyms: Ethyl 2-aminothiazole-5-carboxylate

E0907-1G E0907-5G
TCE0907-1GEA 53.72 CAD
TCE0907-1G TCE0907-5G
Ethyl-2-aminothiazole-5-carboxylate ≥97.0%
Ethyl-2-aminothiazole-5-carboxylate

CAS Number: 32955-21-8
MDL Number: MFCD00602139
Molecular Formula: C6H8N2O2S
Molecular Weight: 172.20
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 161
Lambda max.: 299 nm (EtOH)Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Aminothiazole-5-carboxylic Acid Ethyl Ester

Formula: C₆H₈N₂O₂S
Melting Pt: 161 °C
MDL Number: MFCD00602139
CAS Number: 32955-21-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=446dafe88ccc67a173a6196a68625819510063c2&vid=446dafe88ccc67a173a6196a68625819510063c2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR