trans-4-Ethylcyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

E0904-25G E0904-5G
TCE0904-25GEA 80.58 CAD
TCE0904-25G TCE0904-5G
trans-4-Ethylcyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
trans-4-Ethylcyclohexanecarboxylic acid

CAS Number: 6833-47-2
MDL Number: MFCD01076323
Molecular Formula: C9H16O2
Molecular Weight: 156.23
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 52

Formula: C₉H₁₆O₂
Melting Pt: 52 °C
MDL Number: MFCD01076323
CAS Number: 6833-47-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=61bbac7ca013e2baeb4a84a3bd6b176831047b24&vid=61bbac7ca013e2baeb4a84a3bd6b176831047b24&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR