(4-Bromophenyl)acetic acid ethyl ester ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 2-(4-bromophenyl)acetate, Ethyl (4-bromophenyl)acetate

E0903-25G E0903-5G
TCE0903-25GEA 93.22 CAD
TCE0903-25G TCE0903-5G
(4-Bromophenyl)acetic acid ethyl ester ≥98.0%
(4-Bromophenyl)acetic acid ethyl ester

CAS Number: 14062-25-0
MDL Number: MFCD00016333
Molecular Formula: C10H11BrO2
Molecular Weight: 243.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 144
Flash Point (°C): 110
Freezing point (°C): 29


Other Product Names / Synonyms: 4-Bromophenylacetic Acid Ethyl Ester

Formula: C₁₀H₁₁BrO₂
MW: 243.1
Boiling Pt: 144 °C
Flash Pt: 110 °C
MDL Number: MFCD00016333
CAS Number: 14062-25-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0185d94de31d82dd389173e0f620eabdc694fd9&vid=a0185d94de31d82dd389173e0f620eabdc694fd9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR