Ethyl-2,3-dihydrobenzo[1,4]dioxine-2-carboxylate ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 1,4-Benzodioxane-2-carboxylate, Ethyl 2,3-dihydrobenzo[1,4]dioxine-2-carboxylate, 1,4-Benzodioxane-2-carboxylic acid ethyl ester

E0902-25G E0902-5G
TCE0902-25GEA 540.36 CAD
TCE0902-25G TCE0902-5G
Ethyl-2,3-dihydrobenzo[1,4]dioxine-2-carboxylate ≥98.0%
Ethyl-2,3-dihydrobenzo[1,4]dioxine-2-carboxylate

CAS Number: 4739-94-0
MDL Number: MFCD01099774
Molecular Formula: C11H12O4
Molecular Weight: 208.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 115
Flash Point (°C): 110
Specific Gravity (20/20): 1.21


Other Product Names / Synonyms: 1,4-Benzodioxane-2-carboxylic Acid Ethyl Ester

Formula: C₁₁H₁₂O₄
MW: 208.2100 g/mol
Boiling Pt: 115 °C
Density: 1.21 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD01099774
CAS Number: 4739-94-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d33666a5974f39a2bd0de30be7dc8bf5e2d9d84d&vid=d33666a5974f39a2bd0de30be7dc8bf5e2d9d84d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR