Ethyl-1-hydroxy-1H-1,2,3-triazole-4-carboxylate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Ethyl 1-Hydroxy-1,2,3-triazole-4-carboxylate, 1-Hydroxy-1H-1,2,3-triazole-4-carboxylic acid Ethyl Ester

E0901-1G E0901-5G
TCE0901-1GEA 132.72 CAD
TCE0901-1G TCE0901-5G
Ethyl-1-hydroxy-1H-1,2,3-triazole-4-carboxylate ≥98.0% (by HPLC, titration analysis)
Ethyl-1-hydroxy-1H-1,2,3-triazole-4-carboxylate

CAS Number: 137156-41-3
MDL Number: MFCD06796746
Molecular Formula: C5H7N3O3
Molecular Weight: 157.13
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 105


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Peptide Synthesis
Other Product Names / Synonyms: 1-Hydroxy-1H-1,2,3-triazole-4-carboxylic Acid Ethyl Ester

Formula: C₅H₇N₃O₃
MW: 157.13 g/mol
Melting Pt: 105 °C
MDL Number: MFCD06796746
CAS Number: 137156-41-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6820f80f2630aa21620430a71189ce831052df3&vid=e6820f80f2630aa21620430a71189ce831052df3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR