Ethyl-2-bromothiazole-4-carboxylate ≥95.0% (by GC)

Supplier: TCI America

E0900-1G E0900-25G E0900-5G
TCE0900-1GEA 53.72 CAD
TCE0900-1G TCE0900-25G TCE0900-5G
Ethyl-2-bromothiazole-4-carboxylate ≥95.0% (by GC)
Ethyl-2-bromothiazole-4-carboxylate

CAS Number: 100367-77-9
MDL Number: MFCD03788564
Molecular Formula: C6H6BrNO2S
Molecular Weight: 236.08
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 154
Melting point (°C): 70
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Bromothiazole-4-carboxylic Acid Ethyl Ester

Formula: C₆H₆BrNO₂S
Boiling Pt: 154 °C
Melting Pt: 70 °C
MDL Number: MFCD03788564
CAS Number: 100367-77-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9a8b0d918fbf6b4dddaaa09d3011973dbc7a0c4&vid=f9a8b0d918fbf6b4dddaaa09d3011973dbc7a0c4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR