Ethyl-2-amino-4-methylthiazole-5-carboxylate ≥98.0%
Supplier: TCI America
Synonyms:
Ethyl 2-amino-4-methylthiazole-5-carboxylate
E0898-1G
E0898-5G
TCE0898-1GEA
42.66
CAD
TCE0898-1G
TCE0898-5G
Ethyl-2-amino-4-methylthiazole-5-carboxylate ≥98.0%
Ethyl 2-amino-4-methylthiazole-5-carboxylate
CAS Number: 7210-76-6
MDL Number: MFCD00123414
Molecular Formula: C7H10N2O2S
Molecular Weight: 186.23
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 178
Other Product Names / Synonyms: 2-Amino-4-methylthiazole-5-carboxylic Acid Ethyl Ester
Formula:
C₇H₁₀N₂O₂S Melting Pt: 178 °C |
MDL Number:
MFCD00123414 CAS Number: 7210-76-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...