Ethyl-4-oxo-3-piperidinecarboxylate hydrochloride ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

E0896-25G E0896-5G
TCE0896-25GEA 613.04 CAD
TCE0896-25G TCE0896-5G
Ethyl-4-oxo-3-piperidinecarboxylate hydrochloride ≥98.0% (by HPLC, total nitrogen)
Ethyl-4-oxo-3-piperidinecarboxylate hydrochloride

CAS Number: 4644-61-5
MDL Number: MFCD00043267
Molecular Formula: C8H13NO3
Molecular Weight: 207.65
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 169


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: 3-Ethoxycarbonyl-4-piperidone Hydrochloride, 4-Oxo-3-piperidinecarboxylic Acid Ethyl Ester Hydrochloride

Formula: C₈H₁₃NO₃
Melting Pt: 169 °C
MDL Number: MFCD00043267
CAS Number: 4644-61-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e7b8748f4074f862d4cb2a62aa1adbed813a455&vid=5e7b8748f4074f862d4cb2a62aa1adbed813a455&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR