(-)-Epicatechin gallate ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
(2R,3R)-2-(3,4-Dihydroxyphenyl)-5,7-dihydroxychroman-3-yl 3,4,5-trihydroxybenzoate, (-)-cis-3,3',4',5,7-Pentahydroxyflavane 3-Gallate
CAS Number: 1257-08-5
MDL Number: MFCD00075936
Molecular Formula: C22H18O10
Molecular Weight: 442.38
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 255
Specific rotation [a]20/D: -177 deg (C=1, EtOH)
Storage Temperature: 0-10°C
Product Categories: Bioscience, Antioxidants, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: (-)-cis-3,3',4',5,7-Pentahydroxyflavane 3-Gallate
Formula:
C₂₂H₁₈O₁₀ MW: 442.38 g/mol Melting Pt: 255 °C |
MDL Number:
MFCD00075936 CAS Number: 1257-08-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...