Ethyl-1-ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylate ≥98.0%

Supplier: TCI America

Synonyms: 1-Ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic Acid Ethyl Ester

E0888-25G E0888-5G
TCE0888-25GEA 290.72 CAD
TCE0888-25G TCE0888-5G
Ethyl-1-ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylate ≥98.0%
Ethyl-1-ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylate

CAS Number: 100501-62-0
MDL Number: MFCD00276022
Molecular Formula: C14H12F3NO3
Molecular Weight: 299.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 207


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Ethyl-6,7,8-trifluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic Acid Ethyl Ester

Formula: C₁₄H₁₂F₃NO₃
MW: 299.25 g/mol
Melting Pt: 207 °C
MDL Number: MFCD00276022
CAS Number: 100501-62-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da4fd5a387f9ba7223ff28a4a7baba10b1207e20&vid=da4fd5a387f9ba7223ff28a4a7baba10b1207e20&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR