5-Ethylpyridine-2,3-dicarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
E0887-25G
E0887-5G
TCE0887-25GEA
181.7
CAD
TCE0887-25G
TCE0887-5G
5-Ethylpyridine-2,3-dicarboxylic acid ≥98.0% (by GC, titration analysis)
5-Ethylpyridine-2,3-dicarboxylic acid
CAS Number: 102268-15-5
MDL Number: MFCD00071733
Molecular Formula: C9H9NO4
Molecular Weight: 195.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 156
Other Product Names / Synonyms: 5-Ethylquinolinic Acid
Formula:
C₉H₉NO₄ Melting Pt: 156 °C |
MDL Number:
MFCD00071733 CAS Number: 102268-15-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...