Ethyl 4-hydroxybenzoate ≥99.0% (by GC, titration analysis) for biochemical research
Supplier: TCI America
E0884-25G
E0884-5G
TCE0884-25GEA
83.12
CAD
TCE0884-25G
TCE0884-5G
Ethyl 4-hydroxybenzoate ≥99.0% (by GC, titration analysis) for biochemical research
Ethyl 4-hydroxybenzoate
[for Biochemical Research]
CAS Number: 120-47-8
MDL Number: MFCD00002353
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Boiling point (°C): 178
Melting point (°C): 117
Product Categories: Bioscience, Preservatives for Biochemical Research
Other Product Names / Synonyms: Ethylparaben, 4-Hydroxybenzoic Acid Ethyl Ester
Formula:
C₉H₁₀O₃ Boiling Pt: 178 °C Melting Pt: 117 °C |
MDL Number:
MFCD00002353 CAS Number: 120-47-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...