1-Ethyl-3-methylimidazolium acetate ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 1-Ethyl-3-methylimidazolium Acetate

E0883-25G E0883-5G
TCE0883-25GEA 469.31 CAD
TCE0883-25G TCE0883-5G
1-Ethyl-3-methylimidazolium acetate ≥95.0% (by HPLC, titration analysis)
1-Ethyl-3-methylimidazolium acetate

CAS Number: 143314-17-4
MDL Number: MFCD06798186
Molecular Formula: C8H14N2O2
Molecular Weight: 170.21
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Liquid
Flash Point (°C): 164
Specific Gravity (20/20): 1.10

Formula: C₈H₁₄N₂O₂
MW: 170.21 g/mol
Density: 1.10 (20 °C)
Flash Pt: 164 °C
MDL Number: MFCD06798186
CAS Number: 143314-17-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7628f35ee32efa726d82275740e0546a5aaea7fc&vid=7628f35ee32efa726d82275740e0546a5aaea7fc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR