Ethyl indol-3-ylacetate ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 3-indoleacetate, Ethyl 2-(1H-indol-3-yl)acetate, Ethyl 3-Indoleacetate, IAA ethyl ester, Indole-3-acetic acid ethyl ester, Ethyl indole-3-acetate, Indol-3-ylacetic acid ethyl ester, 3-Indoleacetic acid ethyl ester

E0878-25G E0878-5G
TCE0878-25GEA 344.44 CAD
TCE0878-25G TCE0878-5G
Ethyl indol-3-ylacetate ≥98.0%
Ethyl indol-3-ylacetate

CAS Number: 778-82-5
MDL Number: MFCD00005635
Molecular Formula: C12H13NO2
Molecular Weight: 203.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 166
Melting point (°C): 44
Flash Point (°C): 113


Product Categories: Bioscience, Auxins
Other Product Names / Synonyms: 3-Indoleacetic Acid Ethyl Ester, IAA Ethyl Ester

Formula: C₁₂H₁₃NO₂
MW: 203.24
Boiling Pt: 166 °C
Melting Pt: 44 °C
Flash Pt: 113 °C
MDL Number: MFCD00005635
CAS Number: 778-82-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d65abe8ab15b3b6bf8906782b647ffaee06716b&vid=7d65abe8ab15b3b6bf8906782b647ffaee06716b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR