2-(Ethoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

E0877-1G E0877-5G
TCE0877-1GEA 39.5 CAD
TCE0877-1G TCE0877-5G
2-(Ethoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
2-(Ethoxycarbonyl)phenylboronic acid

CAS Number: 380430-53-5
MDL Number: MFCD02179453
Molecular Formula: C9H11BO4
Molecular Weight: 193.99
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 138


Other Product Names / Synonyms: 2-(Ethoxycarbonyl)benzeneboronic Acid

Formula: C₉H₁₁BO₄
Melting Pt: 138 °C
MDL Number: MFCD02179453
CAS Number: 380430-53-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d631702da561cd1ddca394e929c31680e2e3b4f&vid=5d631702da561cd1ddca394e929c31680e2e3b4f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR