Ethyl-3-methyl-3-phenylglycidate (mixture of cis and trans isomers) ≥95.0% (by GC)

Supplier: TCI America

E0873-250G E0873-25G
TCE0873-250GEA 91.64 CAD
TCE0873-250G TCE0873-25G
Ethyl-3-methyl-3-phenylglycidate (mixture of cis and trans isomers) ≥95.0% (by GC)
Ethyl 3-methyl-3-phenylglycidate (mixture of cis and trans isomers)

CAS Number: 77-83-8
MDL Number: MFCD00022338
Molecular Formula: C12H14O3
Molecular Weight: 206.24
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 86
Flash Point (°C): 134
Specific Gravity (20/20): 1.09


Other Product Names / Synonyms: 3-Methyl-3-phenylglycidic Acid Ethyl Ester, Ethyl 2,3-Epoxy-3-phenylbutyrate, 2,3-Epoxy-3-phenylbutyric Acid Ethyl Ester

Formula: C₁₂H₁₄O₃
Boiling Pt: 86 °C
Density: 1.09 (20 °C)
Flash Pt: 134 °C
MDL Number: MFCD00022338
CAS Number: 77-83-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c04a5fbc52eae26c984fadeb8757f86e20a4825&vid=6c04a5fbc52eae26c984fadeb8757f86e20a4825&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR