Ethyl-4-pyrazolecarboxylate ≥98.0% (by GC)
Supplier: TCI America
E0871-1G
E0871-5G
TCE0871-1GEA
66.36
CAD
TCE0871-1G
TCE0871-5G
Ethyl-4-pyrazolecarboxylate ≥98.0% (by GC)
Ethyl-4-pyrazolecarboxylate
CAS Number: 37622-90-5
MDL Number: MFCD00010844
Molecular Formula: C6H8N2O2
Molecular Weight: 140.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 140
Melting point (°C): 78
Other Product Names / Synonyms: 4-Pyrazolecarboxylic Acid Ethyl Ester
Formula:
C₆H₈N₂O₂ Boiling Pt: 140 °C Melting Pt: 78 °C |
MDL Number:
MFCD00010844 CAS Number: 37622-90-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...