Ethyl-2,4-dihydroxy-6-methyl-3-pyridinecarboxylate ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 2,4-Dihydroxy-6-methyl-3-pyridinecarboxylate, 2,4-Dihydroxy-6-methyl-3-pyridinecarboxylic acid ethyl ester, Ethyl 2,4-dihydroxy-6-methyl-3-pyridinecarboxylate, Ethyl 2,4-Dihydroxy-6-methylnicotinate, 2,4-Dihydroxy-6-methylnicotinic acid ethyl ester

E0869-1G E0869-5G
TCE0869-1GEA 257.54 CAD
TCE0869-1G TCE0869-5G
Ethyl-2,4-dihydroxy-6-methyl-3-pyridinecarboxylate ≥98.0%
Ethyl-2,4-dihydroxy-6-methyl-3-pyridinecarboxylate

CAS Number: 70254-52-3
MDL Number: MFCD02929397
Molecular Formula: C9H11NO4
Molecular Weight: 197.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 210


Other Product Names / Synonyms: 2,4-Dihydroxy-6-methylnicotinic Acid Ethyl Ester, Ethyl 2,4-Dihydroxy-6-methyl-3-pyridinecarboxylate, 2,4-Dihydroxy-6-methyl-3-pyridinecarboxylic Acid Ethyl Ester

Formula: C₉H₁₁NO₄
MW: 197.1900 g/mol
Melting Pt: 210 °C
MDL Number: MFCD02929397
CAS Number: 70254-52-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0647f7804e97c1ec3f342422dd4020fbca1a3792&vid=0647f7804e97c1ec3f342422dd4020fbca1a3792&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR