4-(Ethoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

E0868-1G E0868-5G
TCE0868-1GEA 55.3 CAD
TCE0868-1G TCE0868-5G
4-(Ethoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
4-(Ethoxycarbonyl)phenylboronic acid

CAS Number: 4334-88-7
MDL Number: MFCD02179441
Molecular Formula: C9H11BO4
Molecular Weight: 193.99
Form: Crystal
Color: White
Melting point (°C): 160


Other Product Names / Synonyms: 4-(Ethoxycarbonyl)benzeneboronic Acid

Formula: C₉H₁₁BO₄
Melting Pt: 160 °C
MDL Number: MFCD02179441
CAS Number: 4334-88-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=08d7ee063fed3e6148eb2455a3861a295d2bbf4a&vid=08d7ee063fed3e6148eb2455a3861a295d2bbf4a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR