Ethyl-5-bromonicotinate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

E0866-25G E0866-5G
TCE0866-25GEA 219.78 CAD
TCE0866-25G TCE0866-5G
Ethyl-5-bromonicotinate ≥98.0% (by GC, titration analysis)
Ethyl 5-bromonicotinate

CAS Number: 20986-40-7
MDL Number: MFCD00040366
Molecular Formula: C8H8BrNO2
Molecular Weight: 230.06
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 92
Melting point (°C): 39
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 5-Bromonicotinic Acid Ethyl Ester, Ethyl 5-Bromo-3-pyridinecarboxylate, 5-Bromo-3-pyridinecarboxylic Acid Ethyl Ester

Formula: C₈H₈BrNO₂
Boiling Pt: 92 °C
Melting Pt: 39 °C
MDL Number: MFCD00040366
CAS Number: 20986-40-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a76e1bc42a8574e616fe4ce01cb5215c52677132&vid=a76e1bc42a8574e616fe4ce01cb5215c52677132&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR