Ethyl-1-methyl-3-piperidinecarboxylate ≥95.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1-Methylnipecotic Acid Ethyl Ester, Ethyl 1-Methylnipecotate, 1-Methyl-3-piperidinecarboxylic Acid Ethyl Ester, Ethyl N-methylpiperidine-3-carboxylate

E0863-25G E0863-5G
TCE0863-25GEA 231.47 CAD
TCE0863-25G TCE0863-5G
Ethyl-1-methyl-3-piperidinecarboxylate ≥95.0% (by GC, titration analysis)
Ethyl-1-methyl-3-piperidinecarboxylate

CAS Number: 5166-67-6
MDL Number: MFCD00006496
Molecular Formula: C9H17NO2
Molecular Weight: 171.24
Purity/Analysis Method: >95.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 96
Flash Point (°C): 68
Specific Gravity (20/20): 0.98


Product Categories: Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: 1-Methylnipecotic Acid Ethyl Ester, Ethyl 1-Methylnipecotate, 1-Methyl-3-piperidinecarboxylic Acid Ethyl Ester

Formula: C₉H₁₇NO₂
MW: 171.24 g/mol
Boiling Pt: 96 °C
Density: 0.98 (20 °C)
Flash Pt: 68 °C
MDL Number: MFCD00006496
CAS Number: 5166-67-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57ff1dafa8cb164d1591cfa575cfbb75249b77f8&vid=57ff1dafa8cb164d1591cfa575cfbb75249b77f8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR